Bacitracin zinc salt, potency min. 60 Units/mg

AC22611-0010 AC22611-0050
103517-734EA 45.4 USD
103517-734 200011-280
Bacitracin zinc salt, potency min. 60 Units/mg
Bacitracin zinc salt
Formula: C₆₆H₁₀₁N₁₇O₁₆SZn
Melting Pt: 250 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD28168035
CAS Number: 1405-89-6
EINECS: 215-787-8

Order Now

Specification Test Results

Appearance White to yellow to beige to light grey Powder
Infrared spectrum Conforms
Loss on drying ≤5.0 %
Potency ≥60 U/mg (On dried substance)
pH 6.0 to 7.5 (10 % solution)
Zinc (Zn) 4.0 to 6.0 % (on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR