D-(+)-Glucurono-6,3-lactone 99+%

Synonyms: D-(+)-Glucuronolactone(R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde

AC22592-5000 AC22592-0250 AC22592-1000
200018-666EA 30.29 USD
200018-666 200043-090 200024-912
D-(+)-Glucurono-6,3-lactone 99+%
D(+)-Glucurono-6,3-lactone
Formula: C₆H₈O₆
MW: 176.12 g/mol
Melting Pt: 170…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00135622
CAS Number: 32449-92-6
EINECS: 251-053-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 170 to 176 °C
Loss on drying ≤0.5 % (105 °C, 3 h)
Solubility (5 % in water) Clear colorless to light yellow
Specific optical rotation +17.0° to +20.0° (20 °C, 589 nm) (c=1, water)
Titration excess NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR