Diphenyl phosphate 99%

Synonyms: Diphenyl hydrogen phosphate

AC22569-0250 AC22569-0050
200012-146EA 138 USD
200012-146 200018-858
Diphenyl phosphate 99%
Diphenyl phosphate
Formula: C₁₂H₁₁O₄P
MW: 250.19 g/mol
Melting Pt: 63…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00003033
CAS Number: 838-85-7
EINECS: 212-657-2

Order Now

Specification Test Results

Appearance White Powder or crystals
Infrared spectrum Conforms
Melting point 62 to 70 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR