L-Glutamic acid 5-benzyl ester 99%

Synonyms: gamma-Benzyl L-glutamateH-Glu(OBzl)-OHγ-Benzyl-L-Glutamate

AC22567-0250 AC22567-0050
200006-570EA 73.3 USD
200006-570 103511-376
L-Glutamic acid 5-benzyl ester 99%
L-Glutamic acid 5-benzyl ester
Formula: C₁₂H₁₅NO₄
MW: 237.3 g/mol
Melting Pt: 162…167 °C
Storage Temperature: Refrigerator
EINECS: 216-826-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +26° to +29° (20 °C, 589 nm) (c=2, 1N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR