(S)-(-)-1,1'-Bi-2-naphthol 99%

Warning

Synonyms: (S)-[1,1'-Binaphthalene]-2,2'-diol(S)-BINOL(S)-(-)-1,1'-Binaphthyl-2,2'-Diol

AC22431-0010 AC22431-0050 AC22431-0250
200015-330EA 39.6 USD
200015-330 200015-332 103511-360
(S)-(-)-1,1'-Bi-2-naphthol 99%
(S)-(-)-1,1'-Bi-2-naphthol
Formula: C₂₀H₁₄O₂
MW: 286.33 g/mol
Melting Pt: 205…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00004068
CAS Number: 18531-99-2
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light grey Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 205 to 211 °C
HPLC ≥98.5 %
Specific optical rotation -33° to -38° (20 °C, 589 nm) (c=1, THF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR