(R)-(+)-1,1'-Bi-2-naphthol 99%
|
Danger
|
Synonyms:
(R)-BINOL
, (R)-(+)-1,1'-Binaphthyl-2,2'-Diol
, (R)-[1,1'-Binaphthalene]-2,2'-diol
AC22430-0050
AC22430-0010
AC22430-2500
AC22430-0250
200039-408EA
22.9
USD
200039-408
200015-300
103527-008
200008-212
(R)-(+)-1,1'-Bi-2-naphthol 99%
(R)-(+)-1,1'-Bi-2-naphthol
Formula:
C₂₀H₁₄O₂ MW: 286.33 g/mol Melting Pt: 205…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004068 CAS Number: 18531-94-7 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to light beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 205 to 211 °C |
HPLC | ≥98.5 % |
Specific optical rotation | +33° to +38° (20 °C, 589 nm) (c=1, THF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...