Dodecacarbonyltriruthenium 99%
Synonyms:
Ruthenium carbonyl
, Triruthenium dodecacarbonyl
AC22400-0010
200008-004EA
113
USD
200008-004
Dodecacarbonyltriruthenium 99%
Dodecacarbonyltriruthenium
Formula:
C₁₂O₁₂Ru₃ MW: 639.33 g/mol Melting Pt: 150 °C Storage Temperature: Ambient |
EINECS:
239-287-4 ADR: 6.1,I |
Specification Test Results
Appearance | Orange Crystals |
Infrared spectrum | Conforms |
Elemental Analysis | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...