Ethyl acetimidate hydrochloride 97%
|
Warning
|
AC22410-0500
200025-328EA
78.6
USD
200025-328
Ethyl acetimidate hydrochloride 97%
Ethyl acetimidate hydrochloride
Appearance: White Crystalline powder
Formula:
C₄H₁₀ClNO Storage Temperature: Ambient |
MDL Number:
MFCD00012572 CAS Number: 2208-07-3 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...