2-Chlorophenethylamine 97%

Danger

AC22284-0100
200021-958EA 164 USD
200021-958
2-Chlorophenethylamine 97%
2-Chlorophenethylamine
Formula: C₈H₁₀ClN
Boiling Pt: 120 °C (15 mmHg)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00008185
CAS Number: 13078-80-3
EINECS: 235-982-1
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5530 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR