Sodium ammonium hydrogen phosphate tetrahydrate 99% for analysis

Synonyms: Sodium ammonium phosphate tetrahydrateAmmonium sodium phosphate tetrahydrate

AC22264-0010
200036-554EA 63.6 USD
200036-554
Sodium ammonium hydrogen phosphate tetrahydrate 99% for analysis
Sodium ammonium hydrogen phosphate tetrahydrate
Formula: NaNH₄HPO₄·4H₂O
MW: 209.07 g/mol
Melting Pt: 80 °C
Density: 1.54 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066970
CAS Number: 7783-13-3

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals
Titration with HCl ≥98.5 %
Chloride (Cl) ≤30 ppm
Nitrate (NO3) ≤20 ppm
Sulfate (SO4) ≤100 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm
Insoluble matter ≤0.01 % (in NH4OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR