Propylamine hydrochloride 99+%

Warning

Synonyms: 1-Aminopropane Hydrochloride

AC22189-0250 AC22189-1000
200020-516EA 60.8 USD
200020-516 200032-700
Propylamine hydrochloride 99+%
Propylamine hydrochloride

Appearance: White to light beige Crystalline powder

Formula: C₃H₉N
MW: 95.57 g/mol
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00012911
CAS Number: 556-53-6

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
Titration with HClO4 ≥99.0 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR