2-Sulfobenzaldehyde sodium salt 90% tech.
Synonyms:
Sodium 2-formylbenzenesulfonate
, Sodium 2-formylbenzenesulphonate
, Sodium 2-Formylbenzenesulfonate
, Sodium 2-Formylbenzenesulphonate
, 2-Formylbenzenesulfonic acid sodium salt
, 2-Formylbenzenesulphonic acid sodium salt
AC21932-5000
200002-634EA
155
USD
200002-634
2-Sulfobenzaldehyde sodium salt 90% tech.
2-Sulfobenzaldehyde sodium salt
Formula:
C₇H₆O₄S MW: 208.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007478 CAS Number: 1008-72-6 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥88 % |
Sodium chloride (NaCl) | ≤3 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...