1,4-Bis(2-hydroxyethoxy)benzene 98+%
Synonyms:
1,4-Di(2-hydroxyethoxy)benzene
, Hydroquinone Di(2-hydroxyethyl) Ether
, Hydroquinone Bis(2-hydroxyethyl) Ether
AC21822-0010
AC21822-1000
200049-658EA
208
USD
200049-658
200031-554
1,4-Bis(2-hydroxyethoxy)benzene 98+%
1,4-Bis(2-hydroxyethoxy)benzene
Appearance: White to off-white Crystalline powder or flakes
Formula:
C₁₀H₁₄O₄ MW: 198.22 g/mol Boiling Pt: 185…200 °C (.3 mmHg) Melting Pt: 104…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002861 CAS Number: 104-38-1 |
Specification Test Results
Appearance | White to almost white Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 100 to 107 °C |
GC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...