4'-Ethoxyacetanilide 97%

Danger

Synonyms: 4-AcetophenetidideAcetphenetidinPhenacetinAcetophenetidinN-(4-Ethoxyphenyl)acetamide

AC21659-0025 AC21659-5000 AC21659-1000
103511-424EA 260 USD
103511-424 200013-860 200013-862
4'-Ethoxyacetanilide 97%
4'-Ethoxyacetanilide
Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Melting Pt: 133…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00009094
CAS Number: 62-44-2
EINECS: 200-533-0
ADR: 3,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 133 to 138 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR