Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) 95%

Danger

Synonyms: Methyl-3,6-endomethylenetetrahydrophthalic AnhydrideMethylnadic AnhydrideNadic Methyl Anhydride

AC21627-0010 AC21627-2500 AC21627-0051
200043-984EA 132 USD
200043-984 102777-548 200031-970
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) 95%
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers)

Appearance: Clear light yellow Viscous liquid

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Boiling Pt: 275 °C (760 mmHg)
Density: 1.23 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00167036
CAS Number: 25134-21-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5060 to 1.5080 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR