Cyclopentadienyltitanium trichloride 99%

Danger

AC21577-0100
200024-864EA 198 USD
200024-864
Cyclopentadienyltitanium trichloride 99%
Cyclopentadienyltitanium trichloride
Formula: C₅Cl₃Ti
MW: 214,28 g/mol
Melting Pt: 170 °C
Storage Temperature: Ambient
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR