Tripalmitin (glycerol tripalmitate) 99%

Synonyms: TripalmitinGlycerol tripalmitate

AC21567-0250 AC21567-0010 AC21567-0050
103517-702EA 43.5 USD
103517-702 200039-072 200033-088
Tripalmitin (glycerol tripalmitate) 99%
Tripalmitin (Glycerol tripalmitate)
Formula: C₅₁H₉₈O₆
MW: 807.34
Boiling Pt: 315 °C
Melting Pt: 65…68 °C
Storage Temperature: Freezer
MDL Number: MFCD00008995
EINECS: 209-098-1

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR