Disodium fumarate 98%

Warning

Synonyms: Sodium fumarateFumaric Acid Disodium SaltFumaric acid disodium salt

AC21553-1000 AC21553-0050 AC21553-0025 AC21553-5000
103511-406EA 425.3 USD
103511-406 200007-670 200026-790 103511-408
Disodium fumarate 98%
Disodium fumarate
Formula: C₄H₂Na₂O₄
MW: 160.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064567
CAS Number: 17013-01-3
EINECS: 241-087-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR