O-Phenyl chlorothioformate 99%
|
Danger
|
Synonyms:
Phenyl chlorothionoformate
, Phenyl chlorothionocarbonate
, O-Phenyl carbonochloridothioate
AC21549-0250
103531-822EA
234
USD
103531-822
O-Phenyl chlorothioformate 99%
O-Phenyl chlorothioformate
Formula:
C₇H₅ClOS MW: 172.63 g/mol Boiling Pt: 81…83 °C (6 mmHg) Density: 1.248 g/cm³ Flash Pt: 81 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004920 CAS Number: 1005-56-7 EINECS: 213-736-4 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5800 to 1.5830 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...