Dimidium bromide 98%

Warning

Synonyms: 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide

AC21534-0010 AC21534-2500
200006-340EA 113.89 USD
200006-340 103511-396
Dimidium bromide 98%
Dimidium bromide
Formula: C₂₀H₁₈BrN₃
MW: 380.29 g/mol
Melting Pt: 230 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011757
CAS Number: 518-67-2
ADR: 6.1,I
https://us.vwr.com/stibo/web/std.lang.all/14/12/2301412.jpg

Order Now

Specification Test Results

Appearance Dark red to purple Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 % (on dry substance)
Water ≤5 % (K.F.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR