4-Amino-2-mercaptopyrimidine 97%

Warning

Synonyms: 4-Amino-2-sulfanylpyrimidine4-Aminopyrimidine-2-thiol4-Amino-2-mercaptopyrimidine4-Amino-2-sulphanylpyrimidine

AC21477-0010 AC21477-0050
200047-720EA 188 USD
200047-720 200023-558
4-Amino-2-mercaptopyrimidine 97%
4-Amino-2-mercaptopyrimidine
Formula: C₄H₅N₃S
MW: 127.1676 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006075
CAS Number: 333-49-3

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR