Methyl-3-aminothiophene-2-carboxylate 99%

Warning

Synonyms: Methyl 3-amino-2-thiophenecarboxylate

AC21430-0500 AC21430-0100 AC21430-2500
200013-230EA 60.4 USD
200013-230 103511-352 200006-962
Methyl-3-aminothiophene-2-carboxylate 99%
Methyl 3-aminothiophene-2-carboxylate
Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Boiling Pt: 100…102 °C (.1 mmHg)
Melting Pt: 66…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00009765
CAS Number: 22288-78-4
EINECS: 244-895-8

Order Now

Specification Test Results

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 64 to 66 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR