Glycyrrhizic acid monoammonium salt 98+%

Synonyms: Glycyrrhizin, ammonium saltMonoammonium glycyrrhizinateGlycyrrhizin monoammonium salt

AC21375-1000 AC21375-0250 AC21375-0100
103518-676EA 42.4 USD
103518-676 200025-486 200025-484
Glycyrrhizic acid monoammonium salt 98+%
Glycyrrhizic acid monoammonium salt
Formula: C₄₂H₆₅NO₁₆
MW: 839.97 g/mol
Melting Pt: 209 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00167400
CAS Number: 53956-04-0

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤6.0 % (K.F.)
Heavy metals ≤20 ppm
Sulfated ash ≤0.2 %
Related substances ≤10.0 % 18-alpha-glycyrrhizic acid (HPLC)
Specific optical rotation +49.0° to +54.0° (20 °C, 589 nm) (c=1, EtOH 50% v/v) (on anhydrous substance)
Titration with HClO4 98.0 to 102.0 % (on anhydrous substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR