Copper(I) bromide dimethyl sulfide complex 99%

AC21318-0250
103518-672EA 136 USD
103518-672
Copper(I) bromide dimethyl sulfide complex 99%
Copper (I) bromide dimethyl sulphide complex
Formula: C₂H₆BrCuS
MW: 205.58
Melting Pt: 132 °C (dec.)
Storage Temperature: Ambient
EINECS: 259-292-5

Order Now

Specification Test Results

Appearance Off-white to beige to green to grey Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Iodometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR