2,4-Diaminophenol dihydrochloride 98%

Danger

Synonyms: 4-Hydroxy-1,3-Phenylenediammonium Dichloride

AC21284-0250 AC21284-1000
102775-798EA 129 USD
102775-798 200008-948
2,4-Diaminophenol dihydrochloride 98%
2,4-Diaminophenol dihydrochloride
Formula: C₆H₁₀Cl₂N₂O
Melting Pt: 222 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012979
CAS Number: 137-09-7
EINECS: 205-279-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to grey-green Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR