Cetyldimethylethylammonium bromide 99%

Warning

Synonyms: Ethylhexadecyldimethylammonium BromideEthylhexadecyldimethylammonium bromideEthyl(1-hexadecyl)dimethylammonium bromideDimethylethylhexadecylammonium bromide

AC21187-5000 AC21187-0050 AC21187-1000
200042-862EA 15.9 USD
200042-862 200024-718 200030-680
Cetyldimethylethylammonium bromide 99%
Cetyldimethylethylammonium bromide
Formula: C₂₀H₄₄BrN
MW: 378.4741 g/mol
Melting Pt: 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011774
CAS Number: 124-03-8
EINECS: 204-672-8
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR