Thioflavin T

Synonyms: Basic Yellow 1Thioflavine T2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chlorideSetoflavine TCI 49005

AC21176-0250 AC21176-1000 AC21176-0050
103509-804EA 241 USD
103509-804 103509-802 200014-804
Thioflavin T
Thioflavin T
Formula: C₁₇H₁₉ClN₂S
MW: 318.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011944
CAS Number: 2390-54-7

Order Now

Specification Test Results

Appearance Yellow to golden to orange-brown Powder
Infrared spectrum Conforms
Solubility (1 mg/ml in methanol) Clear to slightly hazy yellow to yellow-green
UV Lambda max 409 to 415 nm (10 mg/l in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR