Benzoyl peroxide 75% remainder water

Danger

Synonyms: Dibenzoyl peroxide

AC21178-0050 AC21178-0010
200057-156EA 196 USD
200057-156 200057-152
Benzoyl peroxide 75% remainder water
Benzoyl peroxide
Formula: C₁₄H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00003071
CAS Number: 94-36-0
UN: 3104
ADR: 5.2,

Order Now

Specification Test Results

Appearance White Moist granular powder
Titration Iodometric 73 to 77 %
Impurity ≤0.25 % Inorganic and organic hydrolizable chloride

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR