D-(-)-Diisopropyl tartrate 99%

Synonyms: (2S,3S)-(-)-Diisopropyl tartrate(2S,3S)-Diisopropyl 2,3-dihydroxysuccinateD-(-)-Tartaric acid diisopropyl ester

AC21118-0500 AC21118-0100
200015-206EA 26.8 USD
200015-206 200008-112
D-(-)-Diisopropyl tartrate 99%
D(-)-Diisopropyl tartrate
Formula: C₁₀H₁₈O₆
MW: 234.25 g/mol
Flash Pt: 110 °C
Storage Temperature: Ambient
EINECS: 263-771-4

Order Now

Specification Test Results

Appearance Clear colorless Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20 °C, 589 nm)
GC ≥98.5 %
Specific optical rotation -16° to -18.5° (20 °C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR