Deuterium chloride 1M in D₂O 99.8 atom % D, AcroSeal™ for NMR spectroscopy

Warning

AC21060-1000
102776-398EA 222 USD
102776-398
Deuterium chloride 1M in D₂O 99.8 atom % D, AcroSeal™ for NMR spectroscopy
Deuterium chloride
Formula: DCl
MW: 37.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044642
CAS Number: 7698-05-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless Liquid
D-Enrichment ≥99.75 %
Strength 0.95 to 1.05 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR