Silver perchlorate hydrate 99%

Danger

AC21021-1000
102777-186EA 368 USD
102777-186
Silver perchlorate hydrate 99%
Silver perchlorate hydrate
Formula: AgClO₄·xH₂O
MW: 207.32 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00149128
CAS Number: 14242-05-8
UN: 1481
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Argentometric ≥98.5 % (on dry substance)
Water ≤10 % (K.F.)
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤20 ppm
Insoluble matter ≤0.005 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR