(1S)-(-)-Camphanic chloride 98%

Danger

Synonyms: (1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride(-)-Camphanic acid chloride

AC20989-0050 AC20989-0250 AC20989-0010
200033-196EA 127 USD
200033-196 102777-646 200008-110
(1S)-(-)-Camphanic chloride 98%
(1S)-(-)-Camphanic chloride
Formula: C₁₀H₁₃ClO₃
MW: 216.67 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135626
CAS Number: 39637-74-6
EINECS: 254-552-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige to grey Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 67 to 71 °C
Specific optical rotation -17° to -25° (20 °C, 589 nm) (c=3.5, CHCl3)
Titration RCOCl ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR