Isobutylmagnesium chloride 2 M in diethyl ether, AcroSeal™

Danger

AC20952-8000
102776-862EA 399.6 USD
102776-862
Isobutylmagnesium chloride 2 M in diethyl ether, AcroSeal™
Isobutylmagnesium chloride

Appearance: Grey to yellow-brown Solution

Formula: C₄H₉ClMg
Density: 0.83 g/cm³
Flash Pt: –040 °C
Storage Temperature: Ambient
MDL Number: MFCD00000471
CAS Number: 5674-02-2
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Grey to yellow-brown Solution
Titration Grignard 2 M Exact strength on the certificate of analysis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR