Sodium 3-nitrobenzenesulfonate 99%
|
Warning
|
Synonyms:
Sodium 3-nitrobenzenesulphonate
, 3-Nitrobenzenesulfonic acid, sodium salt
, 3-Nitrobenzenesulphonic acid, sodium salt
AC20912-0010
200023-400EA
70
USD
200023-400
Sodium 3-nitrobenzenesulfonate 99%
Sodium 3-nitrobenzenesulphonate
Formula:
C₆H₄NNaO₅S MW: 225.16 g/mol Melting Pt: 350 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007490 CAS Number: 127-68-4 EINECS: 204-857-3 |
Specification Test Results
Appearance | Light yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
pH | 6 to 10 (1 % in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...