2-Bromobenzylamine hydrochloride 97%

Warning

AC20909-0250 AC20909-0050
200046-712EA 267 USD
200046-712 200022-486
2-Bromobenzylamine hydrochloride 97%
2-Bromobenzylamine hydrochloride
Formula: C₇H₉BrClN
Melting Pt: 225…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012853
CAS Number: 5465-63-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 229 to 236 °C
Titration Argentometric ≥96.0 %
Water ≤0.5 % (K.F.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR