Chlorohydroquinone 90% tech.

Danger

Synonyms: 2-Chlorobenzene-1,4-diol2-Chlorohydroquinone

AC20822-0050 AC20822-1000
200030-704EA 21.1 USD
200030-704 200018-490
Chlorohydroquinone 90% tech.
Chlorohydroquinone
Formula: C₆H₅ClO₂
MW: 144.56 g/mol
Melting Pt: 101…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002341
CAS Number: 615-67-8
EINECS: 210-442-8
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Grey to light brown Crystalline powder
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR