2-Chloro-3,5-dinitropyridine 98+%

Danger

AC20819-0250 AC20819-0010 AC20819-0050
102775-532EA 109 USD
102775-532 103531-782 103531-784
2-Chloro-3,5-dinitropyridine 98+%
2-Chloro-3,5-dinitropyridine
Formula: C₅H₂ClN₃O₄
MW: 203.54 g/mol
Melting Pt: 63…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006233
CAS Number: 2578-45-2
EINECS: 219-937-3
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Yellow Powder or crystals
Infrared spectrum Conforms
Melting point 61 to 66 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR