Methotrexate hydrate 99%

Danger

Synonyms: Antifolan hydrateMTX hydrate(+)-Amethopterin hydrateL-4-Amino-N10-methylpteroylglutamic acid hydrate(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzoyl)-L-glutamic acid hydrate(1:x)L-Amethopterin hydrate4-Amino-10-methylfolic acid hydrateMethylaminopterin hydrateL(+)-Amethopterin hydrate

AC20810-0250 AC20810-1000 AC20810-5000
102777-244EA 405 USD
102777-244 200019-452 200006-772
Methotrexate hydrate 99%
Methotrexate hydrate
Formula: C₂₀H₂₂N₈O₅·2H₂O
MW: 454.44 g/mol (anhy)
Melting Pt: 195 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00150847
CAS Number: 133073-73-1
EINECS: 200-413-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Water ≤13 % (K.F.)
HPLC ≥98.5 %
Specific optical rotation +19° to +24° (20 °C, 589 nm) (c=1, 14 g/l Na2CO3) (on anhydrous substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR