Retinoic acid 97%

Warning

Synonyms: (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acidall-trans-Retinoic acidVitamin A acidATRATretinoin

AC20734-1000 AC20734-0050
200002-412EA 55.83 USD
200002-412 103511-212
Retinoic acid 97%
Retinoic acid
Formula: C₂₀H₂₈O₂
MW: 300.44 g/mol
Melting Pt: 179…184 °C
Storage Temperature: Freezer
MDL Number: MFCD00001551
CAS Number: 302-79-4
EINECS: 206-129-0

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 184 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR