Papaverine hydrochloride 99%

Danger

AC20722-1000 AC20722-0050 AC20722-0250
200013-890EA 44.4 USD
200013-890 102776-684 102775-596
Papaverine hydrochloride 99%
Papaverine hydrochloride

Appearance: White to beige Crystalline powder

Formula: C₂₀H₂₁NO₄·HCl
Melting Pt: 226 °C
Storage Temperature: Ambient
MDL Number: MFCD00012745
CAS Number: 61-25-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR