Lithium tetrahydroborate 95%

Danger

Synonyms: Lithium borohydrideLithium tetrahydridoborate

AC20681-0100 AC20681-0500
102777-302EA 460 USD
102777-302 102775-780
Lithium tetrahydroborate 95%
Lithium tetrahydroborate
Formula: H₄BLi
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: 280 °C
Density: 0.66 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011088
CAS Number: 16949-15-8
EINECS: 241-021-7
UN: 1413
ADR: 4.3,I

Order Now

Specification Test Results

Appearance White Powder
Assay ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR