Diethylzinc 15% (w/w) 1 M in hexane, AcroSeal™

Danger

AC20551-1001 AC20551-8000
103526-594EA 112 USD
103526-594 102777-112
Diethylzinc 15% (w/w) 1 M in hexane, AcroSeal™
Diethylzinc
Formula: C₄H₁₀Zn
Melting Pt: 39…28 °C
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009021
CAS Number: 557-20-0
EINECS: 209-161-3
UN: 3394
ADR: 4.2,

Order Now

Specification Test Results

Appearance Clear to hazy colorless to yellow to grey-brown Solution
Titration Complexometric 0.85 to 1.00 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR