Ethyl (2-amino-4-thiazolyl)acetate 99%

Synonyms: Ethyl 2-(2-aminothiazol-4-yl)acetate(2-Amino-4-thiazolyl)acetic Acid Ethyl EsterEthyl 2-amino-4-thiazoleacetate

AC20530-0050
200012-348EA 22.8 USD
200012-348
Ethyl (2-amino-4-thiazolyl)acetate 99%
Ethyl (2-amino-4-thiazolyl)acetate
Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Melting Pt: 91…95 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00005330
CAS Number: 53266-94-7

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 91 to 95 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR