2-Hydroxyiminopropiophenone 99%
Synonyms:
1-Phenyl-1,2-propanedione-2-oxime
, 2-Isonitrosopropiophenone
, 1-Phenyl-1,2-propanedione 2-Oxime
, 2-Hydroxyiminopropiophenone
AC20502-0100
AC20502-0250
200004-798EA
63.1
USD
200004-798
200063-486
2-Hydroxyiminopropiophenone 99%
2-Hydroxyiminopropiophenone
Formula:
C₉H₉NO₂ MW: 163.18 g/mol Melting Pt: 111…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002115 CAS Number: 119-51-7 EINECS: 204-329-2 |
Specification Test Results
Appearance | White to light yellow Crystals |
Infrared spectrum | Conforms |
Melting point | 111 to 115 °C |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...