Iminodiacetic acid 98+%

Warning

Synonyms: 2,2'-Azanediyldiacetic acid

AC20497-0250 AC20497-5000
200006-688EA 55.1 USD
200006-688 200019-352
Iminodiacetic acid 98+%
Iminodiacetic acid
Formula: C₄H₇NO₄
MW: 133.10 g/mol
Melting Pt: 243 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004280
CAS Number: 142-73-4
EINECS: 205-555-4

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR