Sodium-D(+)-glucuronate monohydrate 99%

Synonyms: D(+)-Glucuronic acid, sodium salt, monohydrateD-Glucuronic acid sodium salt hydrate

AC20457-1000 AC20457-0250
103516-712EA 190 USD
103516-712 200006-232
Sodium-D(+)-glucuronate monohydrate 99%
Sodium D(+)-glucuronate monohydrate
Formula: C₆H₁₀O₇·1H₂O
MW: 234.14 g/mol (216.14 anhy)
Melting Pt: 134 …140 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151051
CAS Number: 14984-34-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water 6.7 to 8.7 % (K.F.)
Specific optical rotation +20° to +22° (20 °C, 589 nm) (c=2, H2O)
Specific optical rotation after 2 h
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR