Dicoumarol 99%

Danger

Synonyms: Bishydroxycoumarin3,3'-Methylenebis(4-hydroxycoumarin)Dicoumarin

AC20412-0050 AC20412-0250
200018-754EA 87.6 USD
200018-754 200030-926
Dicoumarol 99%
Dicoumarol

Appearance: White Crystalline powder

Formula: C₁₉H₁₂O₆
MW: 336.30 g/mol
Melting Pt: 290…292 °C
Storage Temperature: Ambient
MDL Number: MFCD00006857
CAS Number: 66-76-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with TBAH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR