4-Nitroquinoline-N-oxide 98%

Danger

Synonyms: 4-Nitroquinoline 1-oxide

AC20379-2500 AC20379-0050 AC20379-0010
102777-172EA 389 USD
102777-172 200023-968 200017-680
4-Nitroquinoline-N-oxide 98%
4-Nitroquinoline N-oxide
Formula: C₉H₆N₂O₃
MW: 190.1555 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00006738
CAS Number: 56-57-5
EINECS: 200-281-1
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Ochre to brown Crystalline solid
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR