(-)-Verbenone 94%

Warning

Synonyms: (1S,5S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one(-)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one(1S)-(-)-Verbenone(-)-2-Pinen-4-one

AC20363-0100 AC20363-0500
200019-444EA 163 USD
200019-444 200019-442
(-)-Verbenone 94%
(-)-Verbenone

Appearance: Clear colorless to yellow Liquid

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 227…228 °C (760 mmHg)
Density: 0.97 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00001343
CAS Number: 1196-01-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4970 (20 °C, 589 nm)
GC ≥92 %
Specific gravity (25 °C) 0.96 to 0.98
Specific optical rotation -150° to -250° (20 °C, 589 nm) (C=4, CHCl3) ,reported only for information

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR