Vincristine sulfate 95%

Danger

AC20344-0050
200039-140EA 169 USD
200039-140
Vincristine sulfate 95%
Vincristine sulphate
Formula: C₄₆H₅₈N₄O₁₄S
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00084729
CAS Number: 2068-78-2
EINECS: 218-190-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤12 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR